ENA390552486 Enamine

5-(2H-1,3-benzodioxol-5-yl)-1-methyl-1H-pyrazole-3-carboxylic acid

  • MDL number MFCD14531179
  • Cas Number : 1216231-15-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 214...216
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-127101

Safety Information

msds https://www.enaminestore.com/catalog/EN300-127101
Availability details for
Quantity
  • Availability
  • Add to Cart