ENA390552488 Enamine

1-methyl-2-nitro-1H-imidazole-5-carboxylic acid

  • MDL number MFCD19443277
  • Cas Number : 50700-55-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-127103

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-127103
Availability details for
Quantity
  • Availability
  • Add to Cart