ENA390552489 Enamine

5-methyl-1-(pentan-3-yl)-1H-pyrazole-3-carboxylic acid

  • MDL number MFCD24369094
  • Cas Number : 1432680-78-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 110...112
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-124353

Safety Information

msds https://www.enaminestore.com/catalog/EN300-124353
Availability details for
Quantity
  • Availability
  • Add to Cart