ENA390552499 Enamine

3-(3-methylphenyl)-1,2,4-oxadiazole-5-thiol

  • MDL number MFCD17259802
  • Cas Number : 1343700-58-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 119...121
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-127119

Safety Information

msds https://www.enaminestore.com/catalog/EN300-127119
Availability details for
Quantity
  • Availability
  • Add to Cart