ENA390552502 Enamine

7,9-difluoro-2,3,4,5-tetrahydro-1-benzoxepin-5-one

  • MDL number MFCD11207533
  • Cas Number : 127557-05-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-109272

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-109272
Availability details for
Quantity
  • Availability
  • Add to Cart