ENA390552506 Enamine

1-(4-aminopyridin-3-yl)propan-1-one

  • MDL number MFCD19649485
  • Cas Number : 1341530-36-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-127128

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-127128
Availability details for
Quantity
  • Availability
  • Add to Cart