ENA390552512 Enamine

3-(benzyloxy)-2,2-dimethylcyclobutan-1-one

  • MDL number MFCD21755679
  • Cas Number : 2063-92-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-127143

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-127143
Availability details for
Quantity
  • Availability
  • Add to Cart