ENA390552514 Enamine

3,5-dichloro-2-(chloromethyl)pyridine

  • MDL number MFCD11558921
  • Cas Number : 1076197-56-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-127145

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-127145
Availability details for
Quantity
  • Availability
  • Add to Cart