ENA390552515 Enamine

3-(methoxycarbonyl)-1,2-oxazole-5-carboxylic acid

  • MDL number MFCD20654390
  • Cas Number : 145441-18-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 136-138
Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-127146

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-127146
Availability details for
Quantity
  • Availability
  • Add to Cart