ENA390552523 Enamine

3-carbamoyl-1,2-oxazole-5-carboxylic acid

  • MDL number MFCD18821846
  • Cas Number : 251912-77-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-127156

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-127156
Availability details for
Quantity
  • Availability
  • Add to Cart