ENA390552524 Enamine

(2-ethyl-1,3-thiazol-5-yl)methanol

  • MDL number MFCD12773970
  • Cas Number : 56012-33-0
  • Molecular Weight : 143.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-127157

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-127157
Availability details for
Quantity
  • Availability
  • Add to Cart