ENA390552525 Enamine

2-[chloro(phenyl)methyl]-5-(2-cyclopentylethyl)-1,3,4-oxadiazole

  • MDL number MFCD13663744
  • Cas Number : 1096913-02-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form oil
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-127158

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-127158
Availability details for
Quantity
  • Availability
  • Add to Cart