ENA390552526 Enamine

1H,2H,3H-pyrrolo[2,3-c]pyridine dihydrochloride

  • MDL number MFCD24369173
  • Cas Number : 1432681-13-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-127161

Safety Information

msds https://www.enaminestore.com/catalog/EN300-127161
Availability details for
Quantity
  • Availability
  • Add to Cart