ENA390552543 Enamine

4-(5-Chloro-thiophen-2-yl)-thiazol-2-ylamine

  • MDL number MFCD01312979
  • Cas Number : 123971-45-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 98...100
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-02768

Safety Information

msds https://www.enaminestore.com/catalog/EN300-02768
Availability details for
Quantity
  • Availability
  • Add to Cart