ENA390552547 Enamine

2-(4-chloro-2-nitrophenyl)ethan-1-amine

  • MDL number MFCD16794009
  • Cas Number : 807640-64-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-127245

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-127245
Availability details for
Quantity
  • Availability
  • Add to Cart