ENA390552549 Enamine

3,5-dichloro-4-hydrazinylbenzoic acid

  • MDL number MFCD07785889
  • Cas Number : 887577-44-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-125151

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-125151
Availability details for
Quantity
  • Availability
  • Add to Cart