ENA390552551 Enamine

N-benzyl-2,2,5,5-tetramethyloxolan-3-amine

  • MDL number MFCD22893359
  • Cas Number : 1432679-03-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-127249

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-127249
Availability details for
Quantity
  • Availability
  • Add to Cart