ENA390552557 Enamine

1-(4-chlorophenyl)-5-methyl-1H-pyrazole-3-carboxylic acid

  • MDL number MFCD08443791
  • Cas Number : 126129-22-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-127262

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-127262
Availability details for
Quantity
  • Availability
  • Add to Cart