ENA390552559 Enamine

2-(butan-2-ylsulfanyl)-5-nitrobenzoic acid

  • MDL number MFCD11136032
  • Cas Number : 1019562-00-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-127264

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-127264
Availability details for
Quantity
  • Availability
  • Add to Cart