ENA390552561 Enamine

2-(butylsulfanyl)-5-nitrobenzoic acid

  • MDL number MFCD12643436
  • Cas Number : 154072-75-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-127266

Safety Information

msds https://www.enaminestore.com/catalog/EN300-127266
Availability details for
Quantity
  • Availability
  • Add to Cart