ENA390552562 Enamine

6-(methylamino)-5-nitropyridine-3-carboxylic acid

  • MDL number MFCD21911436
  • Cas Number : 1218943-22-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-127268

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-127268
Availability details for
Quantity
  • Availability
  • Add to Cart