ENA390552566 Enamine

6-amino-5-nitropyridine-2-carboxylic acid

  • MDL number MFCD16556246
  • Cas Number : 84487-06-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-127274

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-127274
Availability details for
Quantity
  • Availability
  • Add to Cart