ENA390552575 Enamine

6-amino-4-methyl-1H-2,3-benzoxazin-1-one

  • MDL number MFCD09878461
  • Cas Number : 217196-94-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-127302

Safety Information

msds https://www.enaminestore.com/catalog/EN300-127302
Availability details for
Quantity
  • Availability
  • Add to Cart