ENA390552583 Enamine

1-{[(2-bromophenyl)methylidene]amino}guanidine

  • MDL number MFCD00465946
  • Cas Number : 170996-63-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-125622

Safety Information

msds https://www.enaminestore.com/catalog/EN300-125622
Availability details for
Quantity
  • Availability
  • Add to Cart