ENA390552586 Enamine

tert-butyl N-(2-methyl-1-oxopropan-2-yl)carbamate

  • MDL number MFCD09751786
  • Cas Number : 109608-77-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-127320

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-127320
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart