ENA390552588 Enamine

ethyl 4-chloro-1,3-dimethyl-1H-pyrazolo[3,4-b]pyridine-5-carboxylate

  • MDL number MFCD00067954
  • Cas Number : 20481-15-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-127322

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-127322
Availability details for
Quantity
  • Availability
  • Add to Cart