ENA390552589 Enamine

tert-butyl N-{[(3-bromophenyl)carbamoyl]methyl}carbamate

  • MDL number MFCD24369191
  • Cas Number : 1387533-90-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-127323

Safety Information

msds https://www.enaminestore.com/catalog/EN300-127323
Availability details for
Quantity
  • Availability
  • Add to Cart