ENA390552591 Enamine

6-(3-ethyl-1,2,4-oxadiazol-5-yl)-2,3,4,5-tetrahydropyridazin-3-one

  • MDL number MFCD18916543
  • Cas Number : 1338682-76-0
  • Molecular Weight : 194.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-127326

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-127326
Availability details for
Quantity
  • Availability
  • Add to Cart