ENA390552598 Enamine

7-methylquinoline-8-sulfonyl chloride

  • MDL number MFCD18791270
  • Cas Number : 17999-75-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 149...151
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-127353

Safety Information

signalword Danger
msds https://www.enaminestore.com/catalog/EN300-127353
pictograms GHS05,GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart