ENA390552599 Enamine

4-(aminomethyl)-N-cyclopropylpyridin-2-amine

  • MDL number MFCD16673900
  • Cas Number : 1247702-69-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 39...41
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-127354

Safety Information

msds https://www.enaminestore.com/catalog/EN300-127354
Availability details for
Quantity
  • Availability
  • Add to Cart