ENA390552601 Enamine

6-chloro-3-ethyl-1,2,3,4-tetrahydropyrimidine-2,4-dione

  • MDL number MFCD19288707
  • Cas Number : 50721-47-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 210...212
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-127356

Safety Information

msds https://www.enaminestore.com/catalog/EN300-127356
Availability details for
Quantity
  • Availability
  • Add to Cart