ENA390552607 Enamine

2-(1H-indol-5-yl)acetic acid

  • MDL number MFCD09837609
  • Cas Number : 34298-84-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 149-150
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-114962

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-114962
Availability details for
Quantity
  • Availability
  • Add to Cart