ENA390552611 Enamine

4,5-difluoro-1H-indole-2-carboxylic acid

  • MDL number MFCD03095188
  • Cas Number : 884494-61-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 209...211
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-127375

Safety Information

msds https://www.enaminestore.com/catalog/EN300-127375
Availability details for
Quantity
  • Availability
  • Add to Cart