ENA390552614 Enamine

N-methyl-N-(propan-2-yl)formamide

  • MDL number MFCD21121123
  • Cas Number : 34855-40-8
  • Molecular Weight : 101.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-127378

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-127378
Availability details for
Quantity
  • Availability
  • Add to Cart