ENA390552615 Enamine

5H,6H,7H-cyclopenta[b]pyridin-3-amine

  • MDL number MFCD09800570
  • Cas Number : 178209-29-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-127390

Safety Information

msds https://www.enaminestore.com/catalog/EN300-127390
Availability details for
Quantity
  • Availability
  • Add to Cart