ENA390552624 Enamine

1-N-[(2-methylphenyl)methyl]benzene-1,2-diamine

  • MDL number MFCD12783973
  • Cas Number : 7191-92-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 71...73
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-125825

Safety Information

msds https://www.enaminestore.com/catalog/EN300-125825
Availability details for
Quantity
  • Availability
  • Add to Cart