ENA390552628 Enamine

4-chloro-2-hydrazinylpyridine

  • MDL number MFCD10000027
  • Cas Number : 364757-36-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-117171

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-117171
Availability details for
Quantity
  • Availability
  • Add to Cart