ENA390552633 Enamine

[(1-phenyl-1H-pyrazol-4-yl)methyl](propan-2-yl)amine

  • MDL number MFCD02656666
  • Cas Number : 1152898-13-9
  • Molecular Weight : 215.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-127414

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-127414
Availability details for
Quantity
  • Availability
  • Add to Cart