ENA390552641 Enamine

4-oxo-4-(3,4,5-trimethoxyphenyl)butanoic acid

  • MDL number MFCD02987265
  • Cas Number : 5101-00-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 109...111
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-127424

Safety Information

msds https://www.enaminestore.com/catalog/EN300-127424
Availability details for
Quantity
  • Availability
  • Add to Cart