ENA390552647 Enamine

3-bromo-5-(1H-imidazol-2-yl)pyridine

  • MDL number MFCD08668549
  • Cas Number : 1211521-74-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-127476

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-127476
Availability details for
Quantity
  • Availability
  • Add to Cart