ENA390552653 Enamine

1-(3-chloro-4-fluorophenyl)pyrrolidin-3-amine

  • MDL number MFCD11894470
  • Cas Number : 1181309-35-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-127482

Safety Information

msds https://www.enaminestore.com/catalog/EN300-127482
Availability details for
Quantity
  • Availability
  • Add to Cart