ENA390552664 Enamine

6-chloro-1H,2H,3H-pyrrolo[2,3-b]pyridine

  • MDL number MFCD18256422
  • Cas Number : 53516-07-7
  • Molecular Weight : 154.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 143-145
Iupac Name 6-chloro-2,3-dihydro-1H-pyrrolo[2,3-b]pyridine
Inchi Code 1S/C7H7ClN2/c8-6-2-1-5-3-4-9-7(5)10-6/h1-2H,3-4H2,(H,9,10)
Inchi Key KBASFRHXGJCOMZ-UHFFFAOYSA-N

Description

Melting Point 143-145
Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-127496

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-127496
pictograms GHS07
hstatements H302,H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart