ENA390552667 Enamine

ethyl 2-(3,4-difluorophenyl)-4-methyl-1,3-thiazole-5-carboxylate

  • MDL number MFCD22581240
  • Cas Number : 317319-19-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 81...83
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-127503

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-127503
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart