ENA390552671 Enamine

3-(pyridin-3-yl)-1,2,4-oxadiazole-5-thiol

  • MDL number MFCD17259796
  • Cas Number : 345631-78-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 179...181
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-126025

Safety Information

msds https://www.enaminestore.com/catalog/EN300-126025
Availability details for
Quantity
  • Availability
  • Add to Cart