ENA390552673 Enamine

6-(tert-butoxy)pyridine-3-carboxylic acid

  • MDL number MFCD11196487
  • Cas Number : 1039902-71-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-127510

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-127510
Availability details for
Quantity
  • Availability
  • Add to Cart