ENA390552674 Enamine

6-fluoro-7-methyl-2,3-dihydro-1H-indole-2,3-dione

  • MDL number MFCD06409073
  • Cas Number : 57817-03-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 204-206
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-127511

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-127511
Availability details for
Quantity
  • Availability
  • Add to Cart