ENA390552680 Enamine

7-chloro-8-(chloromethyl)-3,4-dihydro-2H-1,5-benzodioxepine

  • MDL number MFCD11206253
  • Cas Number : 1094323-67-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 55...57
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-126030

Safety Information

msds https://www.enaminestore.com/catalog/EN300-126030
Availability details for
Quantity
  • Availability
  • Add to Cart