ENA390552681 Enamine

[2-(2-chloro-5-nitropyridin-4-yl)ethenyl]dimethylamine

  • MDL number MFCD00229171
  • Cas Number : 142078-36-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-127521

Safety Information

msds https://www.enaminestore.com/catalog/EN300-127521
Availability details for
Quantity
  • Availability
  • Add to Cart