ENA390552682 Enamine

5-bromo-2-(2,2,2-trifluoroethyl)-1H-1,3-benzodiazole

  • MDL number MFCD20403458
  • Cas Number : 1341801-56-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 139...141
Purity 95.00
Storage temp. -20
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-127523

Safety Information

msds https://www.enaminestore.com/catalog/EN300-127523
Availability details for
Quantity
  • Availability
  • Add to Cart