ENA390552683 Enamine

O-(1-ethylpiperidin-3-yl)hydroxylamine

  • MDL number MFCD16038317
  • Cas Number : 1350738-80-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-127525

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-127525
Availability details for
Quantity
  • Availability
  • Add to Cart